Order Entry
Denmark
ContactUsLinkComponent
CHLORAMPHENICOL, gamma-IRRADIATED. Molecular Biology Reagent Inhibitor of translation on the 50S subunit at the peptidyltransferase step. 1 * 20 mg
Catalog # ICNA0219478720
Supplier:  MP Biomedicals
CHLORAMPHENICOL, gamma-IRRADIATED. Molecular Biology Reagent Inhibitor of translation on the 50S subunit at the peptidyltransferase step. 1 * 20 mg
Catalog # ICNA0219478720
Supplier:  MP Biomedicals
Frequently Bought Together