Order Entry
Denmark
ContactUsLinkComponent
Transparent crate with lid, Material: Polypropylene transparent. Nestable without lid - Stackable with lid. For food. Capacity 20 L. Outside dim 380 x 285 x 295 mm. 1 * 1 items
  LAMA1908
 :  LAMAPLAST
Transparent crate with lid, Material: Polypropylene transparent. Nestable without lid - Stackable with lid. For food. Capacity 20 L. Outside dim 380 x 285 x 295 mm. 1 * 1 items
  LAMA1908
 :  LAMAPLAST
Frequently Bought Together