Order Entry
Denmark
ContactUsLinkComponent
Cleanroom SpillEx - Spill pick-up pad for all water-based liquids (drug manufacturing, body fluids…) - Not suitable for hazardous chemicals 1 * 10 items
  VILE150738
Cleanroom SpillEx - Spill pick-up pad for all water-based liquids (drug manufacturing, body fluids…) - Not suitable for hazardous chemicals 1 * 10 items
  VILE150738
Frequently Bought Together