Biochrom 30+ Amino Acid Analyser. Physiological system comprising instrument, 84 position cooled autosampler, Analytical column with top up resin, prewash column, BioSYS operating software, Ezchrom Elite data manipulation software, chemical kit with 1 * 1 items
Catalog # WAPR80-6000-53
Supplier: Biochrom
Frequently Bought Together