Order Entry
Denmark
ContactUsLinkComponent
Alarm output, zero-voltage - for temperature (±2 °C) and humidity (±5 % RH), accessible via 6-pin DIN socket (max. 24 V - 2.5 A), wi th audible signal that can be switched off 1 * 1 items
Catalog # BIND8012-1001
Supplier:  Binder
Alarm output, zero-voltage - for temperature (±2 °C) and humidity (±5 % RH), accessible via 6-pin DIN socket (max. 24 V - 2.5 A), wi th audible signal that can be switched off 1 * 1 items
Catalog # BIND8012-1001
Supplier:  Binder
Frequently Bought Together