Order Entry
Denmark
ContactUsLinkComponent
An aminopyrimidine that binds a range of tyrosine kinases, including ABL, BTK, FAK, FER, JAK1, SRC, SYK, TEC, TNK1, TYK2, and YES 1 * 1 mg
Catalog # CAYM16718-1
Supplier:  Cayman Chemical
An aminopyrimidine that binds a range of tyrosine kinases, including ABL, BTK, FAK, FER, JAK1, SRC, SYK, TEC, TNK1, TYK2, and YES 1 * 1 mg
Catalog # CAYM16718-1
Supplier:  Cayman Chemical
Frequently Bought Together