A kaempferol glycoside that selectively inhibits RSK2 with an IC50 value of 89 nM (Ki = 1 M) without interfering with upstream acti vators of RSK, including ERK, MEK, EGFR, and PKC 1 * 10 mg
Catalog # CAYM11704-10
Supplier: Cayman Chemical
Frequently Bought Together