Order Entry
Denmark
ContactUsLinkComponent
A potent allosteric inhibitor of PRMT3 (IC50 = 50 nM) with >100-fold selectivity over other methyltransferases and other non-epigene tic targets 1 * 50 mg
  CAYM17017-50
A potent allosteric inhibitor of PRMT3 (IC50 = 50 nM) with >100-fold selectivity over other methyltransferases and other non-epigene tic targets 1 * 50 mg
  CAYM17017-50
Frequently Bought Together