Order Entry
Denmark
ContactUsLinkComponent
A potent allosteric inhibitor of PRMT3 (IC50 = 50 nM) with >100-fold selectivity over other methyltransferases and other non-epigene tic targets 1 * 10 mg
Catalog # CAYM17017-10
Supplier:  Cayman Chemical
A potent allosteric inhibitor of PRMT3 (IC50 = 50 nM) with >100-fold selectivity over other methyltransferases and other non-epigene tic targets 1 * 10 mg
Catalog # CAYM17017-10
Supplier:  Cayman Chemical
Frequently Bought Together