2-(2-Ethoxyethoxy)ethyl acetate
200 L
Catalog # SPCME1023-200LTBL
Supplier: Spectrum Chemical
2-(2-Ethoxyethoxy)ethyl acetate
Catalog # SPCME1023-200LTBL
Supplier: Spectrum Chemical
CAS Number: 112-15-2
Jump to:
Product Details & Documents
Synonyms: Diethylene glycol monoethyl ether acetate
- CAS Number:112-15-2
- Formula:C₈H₁₆O₄
- MDL Number:MFCD00041928
- Molecular Weight:176.21 g/mol
- Boiling Pt:210 °C (1013 hPa)
- Melting Pt:–25 °C
- Density:1.0114 g/cm³ (20 °C)
- EINECS:203-940-1
Specifications
Specification Test Results
- Assay:99% Min.
Specifications
- Pk:200 L



