Order Entry
Denmark
ContactUsLinkComponent
Shelf for 23CF (240bx) ULT freezer, fits Forma 900, 700, Revco ExF, DxF, Thermo Sci TSE, TSD and HERAFreeze HFUB, Isotemp and VWR private label 1 * 1 items
Catalog # 471-1152
Supplier:  VWR Collection
Shelf for 23CF (240bx) ULT freezer, fits Forma 900, 700, Revco ExF, DxF, Thermo Sci TSE, TSD and HERAFreeze HFUB, Isotemp and VWR private label 1 * 1 items
Catalog # 471-1152
Supplier:  VWR Collection
Frequently Bought Together