Order Entry
Denmark
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Unisette™ M406
Turquoise
  SIMPM406-12T
Biopsy processing/embedding cassettes, Unisette™ M406
  SIMPM406-12T
Biopsy processing/embedding cassettes, Unisette™ M406
Biopsy processing/embedding cassettes, Unisette™ M406
Turquoise
2.340,00 kr
1 * 1.000 Items
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
  • Moulded from a special high density polymer
  • Resistant to the chemical action of most histological solvents
  • Colour:
    Turquoise
  • Seq:
    21