Order Entry
Denmark
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Unisette™ M406
Biopsy processing/embedding cassettes, Unisette™ M406
Pink
Catalog # 720-2956
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
Catalog # 720-2956
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
Biopsy processing/embedding cassettes, Unisette™ M406
Pink
Price
2.340,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
  • Moulded from a special high density polymer
  • Resistant to the chemical action of most histological solvents
Specifications
  • Colour:
    Pink
  • Seq:
    13