Biopsy processing/embedding cassettes, Unisette™ M406
Turquoise
Catalog # SIMPM406-12
Supplier: Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
Catalog # SIMPM406-12
Supplier: Simport Scientific
Jump to:
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
- Moulded from a special high density polymer
- Resistant to the chemical action of most histological solvents
Documents
Specifications
- Colour:Turquoise
- Seq:11