Biopsy processing/embedding cassettes, Unisette™ M406
Pink
Catalog # SIMPM406-3
Supplier: Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
Catalog # SIMPM406-3
Supplier: Simport Scientific
Jump to:
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
- Moulded from a special high density polymer
- Resistant to the chemical action of most histological solvents
Specifications
- Colour:Pink
- Seq:02