Order Entry
Denmark
Orders LinkContactUsLinkComponent
Control slides, ISOSLIDE®
Iron control slides
  1.00380.0001
 :   Merck
Control slides, ISOSLIDE®
  1.00380.0001
 :  Merck
Control slides, ISOSLIDE®
Control slides, ISOSLIDE®
Iron control slides
2.400,00 kr
1 * 1 Each
ISOSLIDE® control slides are designed for use with new as well as existing products.
  • Shelf life
  • Prepared exclusively from animal material
  • All slides in a box are from the same batch and are prepared from one block
ISOSLIDE® control slides help optimise and standardise staining results by allowing typical control material to be compared with a laboratory’s own specimens. Typical material is prepared and sliced using standard procedures. One slide is already stained by the Merck staining procedure, while the other 24 are unstained paraffin sections. These slides can be stained using the laboratory’s own method or according to the MERCK protocol. For every pack of ISOSLIDE® control slides an insert sheet referring to the relevant products is available.
  • Description:
    Iron control slides