Order Entry
Denmark
Orders LinkContactUsLinkComponent
Multi-element standards, comprehensive mix B, VHG

Multi-element standards, comprehensive mix B, VHG

Comprehensive mix B standard, 1× 100 ml, 100 mL
Catalog # PROOVHG-SM90C-100
Multi-element standards, comprehensive mix B, VHG
Catalog # PROOVHG-SM90C-100
CAS Number:  
Multi-element standards, comprehensive mix B, VHG

Comprehensive mix B standard, 1× 100 ml, 100 mL

CAS Number:  
Price
6.850,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents

This standard solution is intended for use as a certified reference material and calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), or alternative techniques, such as and flame or furnace atomic absorption spectroscopy (AA or GFAA).

Multi element standards for Ag, Al, As, Ba, Bi, Ca, Cd, Ce, Dy, Er, Eu, Ga, Gd, Ho, La, Lu, Mg, Na, Nd, P, Pb, Pr, Rb, Sc, Se, Sm, Sr, Tb, Th, Tl, Tm, U, Y and Yb at 10 µg/ml in 40% aqua regia, tr. HF.

Certifications: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Comprehensive mix B standard
  • Pk Info:
    1× 100 ml

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.