Caffeine
Caffeine
Katalog # EHERCR11693000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: CN1C(=O)N(C)c2ncn(C)c2C1=O
- Inchi: InChI=1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3
- IUPAC: 1,3,7-trimethylpurine-2,6-dione
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Description:Caffeine
- Cat. No.:EHERCR11693000
- Formula:C₈H₁₀N₄O₂
- Storage conditions:20 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):Caffeine;Theophylline-Ethylenediamine Imp. A (EP);Pentoxifylline Imp. F (EP);Dimenhydrinate Imp. C (EP);Theophylline Imp. A (EP);Theophylline-Ethylenediamine Anhydrous Imp. A (EP);Caffeine (8CI);3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione;1,3,7-Trimethyl-2,6-dioxopurine;1,3,7-Trimethyl-7H-purine-2,6-dione;1,3,7-Trimethylxanthine;7-Methyltheophylline;Alert-Pep;Asia migrine;Cafalgine;Cafeina;Caffedrine;Caffein;Cafipel;DHCplus;Dasin;Diurex;Durvitan;Guaranine;Koffein;Mateina;Methyltheobromine;Midron extra;Miudol;NSC 5036;New Cetamol;No-Doz;Palergot-C;Phensal;Refresh'n;Shape Plus;Stay Alert;Stim;Synalgos;Thein;Theine;Tri-Aqua;Vivarin;Wigraine;Dimenhydrinate Impurity C;Pentoxifylline Impurity F;Theophylline Impurity A;Theophylline-Ethylenediamine Impurity A
- Molecular weight:194,19
- CAS No.:58-08-2
- Shipping conditions:RT
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



