Order Entry
Denmark
Orders LinkContactUsLinkComponent
Universal pathogen DNA kit, Mag-Bind®

Universal pathogen DNA kit, Mag-Bind®

Mag-Bind® Universal Pathogen DNA Kit (4×96), 1 KIT
Catalog # M4029-01
Supplier:   OMEGA BIO-TEK
Universal pathogen DNA kit, Mag-Bind®
Catalog # M4029-01
Supplier:  OMEGA BIO-TEK
CAS Number:  
Universal pathogen DNA kit, Mag-Bind®
Universal pathogen DNA kit, Mag-Bind®

Mag-Bind® Universal Pathogen DNA Kit (4×96), 1 KIT

CAS Number:  
Price
14.660,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

The Mag-Bind® Universal Pathogen DNA 96 Kit allows rapid and reliable isolation of high-quality pathogen and host genomic DNA from tissue samples. Up to a 30 mg tissue sample is recommended and can be processed in less than 60 minutes. The system allows for automation after sample lysis via Hamilton STAR™, Thermo KingFisher™ Flex, Applied Biosystems® MagMAX™, Qiagen Biosprint, and other liquid handling instruments.

The system combines the Mag-Bind® technology with RBB Buffer to eliminate PCR inhibiting compounds within the samples. Purified DNA is suitable for PCR, restriction digestion, and hybridisation applications.There are no organic extractions, thus reducing plastic waste and hands-on time and multiple samples can be processed in parallel.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Mag-Bind® Universal Pathogen DNA Kit (4×96)
  • Prep size:
    Scalable
  • Description:
    Mag-Bind® Universal Pathogen DNA Kit
  • Sample type:
    Stool|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Sample size:
    Variable
  • Cat. No.:
    M4029-01
  • Refine target molecule:
    Viral DNA|Bacterial DNA
  • Target molecule:
    DNA
  • Method / Format:
    Magnetic beads

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.