Order Entry
Denmark
ContactUsLinkComponent
Vial Convenience Pack LVI™, Clear Glass Limited Volume Autosampler Vial with 0.3mL Insert, 9mm Natural ABC Screw Cap & Red PTFE/Silicone/PTFE Septa 1 * 100 items
Catalog # WHEAW225326-03
Supplier:  DWK Life Sciences
Vial Convenience Pack LVI™, Clear Glass Limited Volume Autosampler Vial with 0.3mL Insert, 9mm Natural ABC Screw Cap & Red PTFE/Silicone/PTFE Septa 1 * 100 items
Catalog # WHEAW225326-03
Supplier:  DWK Life Sciences
Frequently Bought Together