Order Entry
Denmark
ContactUsLinkComponent
Stainless steel bottom drip tray, 15 mm rim (may affect the temperature distribution) – cannot be used in connection with option K1 MAX. LOADING 4 KG 1 * 1 items
Catalog # MEMMB29722UF260TS
Supplier:  MEMMERT
Stainless steel bottom drip tray, 15 mm rim (may affect the temperature distribution) – cannot be used in connection with option K1 MAX. LOADING 4 KG 1 * 1 items
Catalog # MEMMB29722UF260TS
Supplier:  MEMMERT
Frequently Bought Together