Order Entry
Denmark
ContactUsLinkComponent
This product is lectin PNA labeled with our superior red fluorescent dye CF568. Lectin PNA is specific for terminal -galactose and binds preferentially to a commonly occurring structure, galactosyl (-1,3) N-acetylgalactosamine. 1 * 1 mg
Catalog # 29061.
Supplier:  Biotium
This product is lectin PNA labeled with our superior red fluorescent dye CF568. Lectin PNA is specific for terminal -galactose and binds preferentially to a commonly occurring structure, galactosyl (-1,3) N-acetylgalactosamine. 1 * 1 mg
Catalog # 29061.
Supplier:  Biotium
Frequently Bought Together