Order Entry
Denmark
Orders LinkContactUsLinkComponent
NMR sample tube washer
NMR sample tube washer
NMR sample tube washer, acc. to Swiegot, complete, to fit laboratory bottles with GL 45 thread
Catalog # WITG2.900.032
Supplier:   witeg Labortechnik
NMR sample tube washer
Catalog # WITG2.900.032
Supplier:  witeg Labortechnik
NMR sample tube washer
NMR sample tube washer
NMR sample tube washer, acc. to Swiegot, complete, to fit laboratory bottles with GL 45 thread
Price
2.640,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Complete with 2 PTFE-adapters to fir 5 cuvets or NMR tubes of 5 and 10 mm diameter in different lengths. Vacuum operated washers allowing the individual washing of 1 to 5 tube, complete with caps and FEPcappilaries (box 1 pce.).
Specifications
  • Description:
    NMR sample tube washer, acc. to Swiegot, complete, to fit laboratory bottles with GL 45 thread
  • Seq:
    02