Cyclohexane ACS
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Hexahydrobenzene, Hexamethylene
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Boiling Pt:81 °C (760 mmHg)
- Melting Pt:6.5 °C
- Density:0.77 g/cm³
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- UN:1145
- ADR:3,II
Documents
ACRO44628
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.02 %
- GC:≥99.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Substance darkened by H2SO4:Passes test
Specifications
- Pack type:Glass bottle
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



