Order Entry
Denmark
Orders LinkContactUsLinkComponent
Pyrogallol ACS

Pyrogallol ACS

Plastic bottle, 2,5 kg
  ACRO418620025
 
 
Pyrogallol ACS
  ACRO418620025
 :  87-66-1
Pyrogallol ACS
Pyrogallol ACS

Plastic bottle, 2,5 kg

 :  87-66-1
 
10.930,00 kr
   
1 * 2,5 kg
 
 :  Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
  •  :87-66-1
  •  :C₆H₆O₃
  •  :MFCD00002192
  •  :126.11 g/mol
  •  :13,08090
  •  :309 °C (1013 hPa)
  •  :131…135 °C
  •  :1,453 g/cm³ (20 °C)
  •  :Ambient
  •  :201-762-9
  •  :2811
  •  :6.1,III

Documents

 

 

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    131.0 to 135.0 °C
  • Residue after ignition:
    ≤0.005 %
  • Chloride (Cl):
    ≤10 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤5 ppm

 

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.