Anthrone 95%, pure
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Anthracen-9(10H)-one
- CAS Number:90-44-8
- Formula:C₁₄H₁₀O
- MDL Number:MFCD00001187
- Molecular Weight:194.23 g/mol
- Boiling Pt:721 °C (760 mmHg)
- Melting Pt:152…157 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:Yellow to beige Crystalline powder or needles
- Infrared spectrum:Conforms
- Melting point:155 to 161 °C
- GC:≥94 %
Specifications
- Pack type:Glass bottle
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



