Pyridine 99+% for analysis
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Pyridine 99+% for analysis
Catalog # ACRO290860025
Supplier: Thermo Fisher Scientific
CAS Number: 110-86-1
Jump to:
Product Details & Documents
Synonyms: Azabenzene, Azine
- CAS Number:110-86-1
- Formula:C₅H₅N
- MDL Number:MFCD00011732
- Molecular Weight:79.1 g/mol
- Boiling Pt:115…116 °C (760 mmHg)
- Melting Pt:–42 °C
- Density:0.978 g/cm³
- Flash Pt:17 °C
- Storage Temperature:Ambient
- UN:1282
- ADR:3,II
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.1 %
- GC:≥99.0 %
- Color scale:≤10 APHA
- Non-volatile matter:≤0.002 %
- Ammonia (NH3):≤0.002 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



