Murexide ACS
Glass bottle, 100 g
: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- :3051-09-0
- :C₈H₈N₆O₆
- :MFCD00012777
- :284.19 g/mol
- :13,06330
- :>300 °C
- :Ambient
- :221-266-6
Documents
- Appearance:Bordeaux to red-purple Powder
- Infrared spectrum:Conforms
- Suitability:(as complexom. indicator) Passes test
- Pack type:Glass bottle
- Pk:100 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



