Murexide ACS
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- CAS Number:3051-09-0
- Formula:C₈H₈N₆O₆
- MDL Number:MFCD00012777
- Molecular Weight:284.19 g/mol
- Merck Index:13,06330
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:221-266-6
Documents
Specifications
Specification Test Results
- Appearance:Bordeaux to red-purple Powder
- Infrared spectrum:Conforms
- Suitability:(as complexom. indicator) Passes test
Specifications
- Pack type:Glass bottle
- Pk:5 g



