Order Entry
Denmark
Orders LinkContactUsLinkComponent
Formamide 99.5% for analysis

Formamide 99.5% for analysis

Plastic drum, 10 L
Catalog # ACRO205820100
Health
Formamide 99.5% for analysis
Catalog # ACRO205820100
CAS Number:  75-12-7
Formamide 99.5% for analysis
Formamide 99.5% for analysis

Plastic drum, 10 L

CAS Number:  75-12-7
Price
3.850,00 kr
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Carbamaldehyde
  • CAS Number:75-12-7
  • Formula:HCONH₂
  • MDL Number:MFCD00007941
  • Molecular Weight:45.04 g/mol
  • Merck Index:13,04261
  • Boiling Pt:210 °C (760 mmHg)
  • Melting Pt:2…3 °C
  • Density:1.133 g/cm³
  • Flash Pt:175 °C
  • Storage Temperature:Ambient
  • EINECS:200-842-0

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 % (Coulometric)
  • Sulfated ash:
    ≤0.005 %
  • GC:
    ≥99.5 %
  • Color scale:
    ≤10 APHA
  • Copper (Cu):
    ≤1 ppm
  • Iron (Fe):
    ≤1 ppm
  • Zinc (Zn):
    ≤1 ppm
  • Heavy metals (as Pb):
    ≤1 ppm

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.