D-(-)-Serine 98%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (R)-(-)-Serine, H-D-Ser-OH
- CAS Number:312-84-5
- Formula:C₃H₇NO₃
- MDL Number:MFCD00004269
- Molecular Weight:105.09 g/mol
- Merck Index:12,08604
- Melting Pt:220 °C (dec.)
- Storage Temperature:Ambient
- EINECS:206-229-4
Documents
ACRO22707
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.3 % (105°C, 3 h)
- Sulfated ash:≤0.1 %
- Specific optical rotation:-13.0° to -16.0° (20°C, 589 nm) (c=10, 2 N HCl)
- Titration with HClO4:97.5 to 102.5 %
- Other amino acids:≤0.5 % (TLC)
Specifications
- Pack type:Glass bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



