Potassium sulphate, pure
Plastic bottle, 1 kg
Supplier: Thermo Fisher Scientific
Potassium sulphate, pure
Catalog # ACRO193810010
Supplier: Thermo Fisher Scientific
CAS Number: 7778-80-5
Jump to:
Product Details & Documents
- CAS Number:7778-80-5
- Formula:K₂SO₄
- MDL Number:MFCD00011388
- Molecular Weight:174.26 g/mol
- Boiling Pt:1689 °C (760 mmHg)
- Melting Pt:1067 °C
- Density:2,66 g/cm³ (20 °C)
- Storage Temperature:Ambient
Documents
ACRO19381
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Loss on drying:≤1 % (130°C)
- pH:5.5 to 8.5
- Titration with HClO4:≥99.0 %
- Arsenic (As):≤0.0003 %
- Lead (Pb):≤0.0002 %
- Mercury (Hg):≤0.0001 %
- Selenium (Se):≤0.0005 %
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



