Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cyclohexane 99.5%, extra pure

Cyclohexane 99.5%, extra pure

Plastic drum, 10 L
Catalog # ACRO210570100
Irritant
Health
Environment
Flammable
Cyclohexane 99.5%, extra pure
Catalog # ACRO210570100
CAS Number:  110-82-7
Cyclohexane 99.5%, extra pure
Cyclohexane 99.5%, extra pure

Plastic drum, 10 L

CAS Number:  110-82-7
Price
3.180,00 kr
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (760 mmHg)
  • Melting Pt:6.5 °C
  • Density:0.77 g/cm³
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.01 %
  • GC:
    ≥99.5 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.005 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.