Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stearic acid 97%

Stearic acid 97%

Fibre drum, 10 kg
Catalog # ACRO174490100
Stearic acid 97%
Catalog # ACRO174490100
CAS Number:  57-11-4
Stearic acid 97%
Stearic acid 97%

Fibre drum, 10 kg

CAS Number:  57-11-4
Price
kr1,660.00
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  Octadecanoic acid, Octadecylic acid
  • CAS Number:57-11-4
  • Formula:C₁₈H₃₆O₂
  • MDL Number:MFCD00002752
  • Molecular Weight:284.48 g/mol
  • Merck Index:13,08882
  • Boiling Pt:361 °C (760 mmHg)
  • Melting Pt:67…69 °C
  • Density:0.84 g/cm³
  • Flash Pt:196 °C
  • Storage Temperature:Ambient
  • EINECS:200-313-4

Documents

Specifications

Specification Test Results

  • Appearance:
    White Granular powder or granules or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    67 to 71 °C
  • GC:
    ≥96.0 %

Specifications

  • Pack type:
    Fibre drum
  • Pk:
    10 kg

Specifications