Squalene 99+%
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene
- CAS Number:111-02-4
- Formula:C₃₀H₅₀
- MDL Number:MFCD00008912
- Molecular Weight:410.73 g/mol
- Merck Index:13,08847
- Boiling Pt:285 °C (25 mmHg)
- Melting Pt:–75 °C
- Density:0.855 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:203-826-1
Documents
ACRO20747
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4945 to 1.4965 (20°C, 589 nm)
- Density:(20°C) 0.845 to 0.865 g/ml
- GC:≥99.0 %
- Acid value:≤1 mg KOH/g
- Iodine value:360 to 400 g I2/100 g
- Peroxide value:≤2 meq/kg
- Saponification value:≤1 mg KOH/g
Specifications
- Pack type:Glass bottle
- Pk:500 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



