Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spermidine 99%

Spermidine 99%

Glass bottle, 100 g
Katalog # ACRO132741000
Ætsende
Leverandør:   Thermo Fisher Scientific
Spermidine 99%
Katalog # ACRO132741000
CAS nummer:  124-20-9
Spermidine 99%
Spermidine 99%

Glass bottle, 100 g

CAS nummer:  124-20-9
Price
14.380,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
  • CAS nummer:124-20-9
  • Formel:C₇H₁₉N₃
  • MDL nummer:MFCD00008229
  • Molecular Weight:145.25 g/mol
  • Merck Index:12,08893
  • Kogende Pt:128…130 °C (14 mmHg)
  • Smeltende Pt:22…25 °C
  • Tæthed:0.925 g/cm³
  • Flash Pt:>110 °C
  • Opbevaringstemperatur:Refrigerator
  • EINECS:204-689-0
  • FN:3259
  • ADR:8,II

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Clear colorless Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4785 to 1.4805 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specifikationer

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifikationer