Spermidine 99%
Glass bottle, 25 g
: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
- :124-20-9
- :C₇H₁₉N₃
- :MFCD00008229
- :145.25 g/mol
- :12,08893
- :128…130 °C (14 mmHg)
- :22…25 °C
- :0.925 g/cm³
- :>110 °C
- :Refrigerator
- :204-689-0
- :3259
- :8,II
Documents
- Appearance:Clear colorless Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.4785 to 1.4805 (20°C, 589 nm)
- GC:≥98.5 %
- Pack type:Glass bottle
- Pk:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



