Order Entry
Denmark
Orders LinkContactUsLinkComponent
Quinoline 96%

Quinoline 96%

Glass bottle, 2,5 L
Catalog # ACRO132190025
Toxic
Health
Environment
Quinoline 96%
Catalog # ACRO132190025
CAS Number:  91-22-5
Quinoline 96%
Quinoline 96%

Glass bottle, 2,5 L

CAS Number:  91-22-5
Price
1.860,00 kr
Log in for availability
1 * 2,5 L
Product Details & Documents
  • CAS Number:91-22-5
  • Formula:C₉H₇N
  • MDL Number:MFCD00006736
  • Molecular Weight:129.16 g/mol
  • Merck Index:13,08160
  • Boiling Pt:237 °C (760 mmHg)
  • Melting Pt:–15 °C
  • Density:1.095 g/cm³
  • Flash Pt:101 °C
  • Storage Temperature:Ambient
  • EINECS:202-051-6
  • UN:2656
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear light yellow to dark brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6230 to 1.6270 (20°C, 589 nm)
  • GC:
    ≥95 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.