Order Entry
Denmark
Orders LinkContactUsLinkComponent
Propyl-4-hydroxybenzoate 99+%

Propyl-4-hydroxybenzoate 99+%

Plastic bottle, 2,5 kg
Catalog # ACRO131590025
Irritant
Propyl-4-hydroxybenzoate 99+%
Catalog # ACRO131590025
CAS Number:  94-13-3
Propyl-4-hydroxybenzoate 99+%
Propyl-4-hydroxybenzoate 99+%

Plastic bottle, 2,5 kg

CAS Number:  94-13-3
Price
3.550,00 kr
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester
  • CAS Number:94-13-3
  • Formula:C₁₀H₁₂O₃
  • MDL Number:MFCD00002354
  • Molecular Weight:180.2 g/mol
  • Merck Index:13,07958
  • Boiling Pt:292 °C (1013 hPa)
  • Melting Pt:95…99 °C
  • Density:1,28 g/cm³ (20 °C)
  • Flash Pt:180 °C
  • Storage Temperature:Ambient
  • EINECS:202-307-7

Documents

Specifications

Specification Test Results

  • Appearance:
    White to almost white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    95 to 99 °C
  • Sulfated ash:
    ≤0.1 %
  • GC:
    ≥99.0 %
  • Appearance of solution:
    (10% in ethanol 96%) clear colorless to

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.