Order Entry
Denmark
Orders LinkContactUsLinkComponent
Thymolphthalein, pure indicator

Thymolphthalein, pure indicator

Glass bottle, 250 g
Catalog # ACRO151462500
Thymolphthalein, pure indicator
Catalog # ACRO151462500
CAS Number:  125-20-2
Thymolphthalein, pure indicator
Thymolphthalein, pure indicator

Glass bottle, 250 g

CAS Number:  125-20-2
Price
4.840,00 kr
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
  • CAS Number:125-20-2
  • Formula:C₂₈H₃₀O₄
  • MDL Number:MFCD00005909
  • Molecular Weight:430.54 g/mol
  • Merck Index:12,09542
  • Melting Pt:251…253 °C
  • Storage Temperature:Ambient
  • EINECS:204-729-7

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 %
  • Sulfated ash:
    ≤0.5 %
  • Visual transition interval:
    from pH 8.8 (colorless) to pH 10.5 (blue)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g

Specifications