Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bromophenol blue, pure indicator

Bromophenol blue, pure indicator

Glass bottle, 100 g
Catalog # ACRO151341000
Bromophenol blue, pure indicator
Catalog # ACRO151341000
CAS Number:  115-39-9
Bromophenol blue, pure indicator
Bromophenol blue, pure indicator

Glass bottle, 100 g

CAS Number:  115-39-9
Price
2.460,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Merck Index:13,01429
  • Melting Pt:279 °C
  • Density:1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-086-2

Documents

Specifications

Specification Test Results

  • Appearance:
    Pink-orange to red Powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.1 % in ethanol) Clear
  • Specific extinction E (1%/1cm):
    ≥1100 (592 nm, in buffer)
  • Visual transition interval:
    from pH 3.0 (yellow) to pH 4.6 (blue)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifications