Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

D(+)-Galactose 99+%

Plastic drum, 5 kg
Catalog # ACRO150610051
D(+)-Galactose 99+%
Catalog # ACRO150610051
CAS Number:  59-23-4
D(+)-Galactose 99+%
D(+)-Galactose 99+%

Plastic drum, 5 kg

CAS Number:  59-23-4
Price
14.050,00 kr
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04356
  • Boiling Pt:527 °C (1013 hPa)
  • Melting Pt:165…168 °C
  • Density:1,5 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:200-416-4
Specifications

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.3 % (K.F.)
  • Sulfated ash:
    ≤0.1 % (on dry substance)
  • Solubility:
    (10 % in water) Clear colorless
  • HPLC:
    ≥99.0 % (ELSD)
  • Specific optical rotation:
    +78° to +81.5° (20°C, 589 nm) (c=10, water, NH3) (after 30 minutes, on dry substance)

Specifications

  • Pakning:
    Plastic drum
  • Pakn:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.