3-Methylpentane 99+%
Glass bottle, 50 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:96-14-0
- Formula:C₆H₁₄
- MDL Number:MFCD00009342
- Molecular Weight:86.18 g/mol
- Boiling Pt:64 °C (760 mmHg)
- Melting Pt:–118 °C
- Density:0.66 g/cm³
- Flash Pt:–32 °C
- Storage Temperature:Ambient
- EINECS:202-481-4
- UN:1208
- ADR:3,II
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3755 to 1.3775 (20°C, 589 nm)
- GC:≥99.0 %
- Assay:≤8 % (Tetracyclin hydrochloride) On dry substance
- Water:≤2.0 %
- Sulfated ash:≤0.5 %
- Identification:Passes test
- pH:2.3 to 3.3 (1 % in water CO2 free)
- Specific optical rotation:-235° to -250° (20°C, 589 nm) (c=0.25, water) (on dry substance)
- Absorbance:≤0.40 (460 nm)
- Heavy metals (as Pb):≤50 ppm
Specifications
- Pack type:Glass bottle
- Pk:50 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



