3-Methylpentane 99+%
Glass bottle, 1 L
- :96-14-0
- :C₆H₁₄
- :MFCD00009342
- :86.18 g/mol
- :64 °C (760 mmHg)
- :–118 °C
- :0.66 g/cm³
- :–32 °C
- :Ambient
- :202-481-4
- :1208
- :3,II
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3755 to 1.3775 (20°C, 589 nm)
- GC:≥99.0 %
- Assay:≤8 % (Tetracyclin hydrochloride) On dry substance
- Water:≤2.0 %
- Sulfated ash:≤0.5 %
- Identification:Passes test
- pH:2.3 to 3.3 (1 % in water CO2 free)
- Specific optical rotation:-235° to -250° (20°C, 589 nm) (c=0.25, water) (on dry substance)
- Absorbance:≤0.40 (460 nm)
- Heavy metals (as Pb):≤50 ppm
- Pack type:Glass bottle
- Pk:1 L



