Eugenol 99%
Glass bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole
- CAS Number:97-53-0
- Formula:C₁₀H₁₂O₂
- MDL Number:MFCD00008654
- Molecular Weight:164.2 g/mol
- Merck Index:13,03931
- Boiling Pt:254 °C (760 mmHg)
- Melting Pt:12…10 °C
- Density:1.06 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:202-589-1
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow-green Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5400 to 1.5420 (20°C, 589 nm)
- GC:≥98.5 %
- Specific gravity:(20°C) 1.064 to 1.070
Specifications
- Pack type:Glass bottle
- Pk:2,5 kg



