Valeric acid 99%
Glass bottle, 100 mL
: Pentanoic acid
- :109-52-4
- :C₅H₁₀O₂
- :MFCD00004413
- :102.13 g/mol
- :12,10042
- :185 °C (760 mmHg)
- :20…18 °C
- :0.94 g/cm³
- :86 °C
- :Ambient
- :203-677-2
- :3265
- :8,II
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4070 to 1.4100 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥98.5 %
- Pack type:Glass bottle
- Pk:100 mL



