Phenol 99%, extra pure
Glass bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene
- CAS Number:108-95-2
- Formula:C₆H₅OH
- MDL Number:MFCD00002143
- Molecular Weight:94.11 g/mol
- Merck Index:12,07390
- Boiling Pt:182 °C (760 mmHg)
- Melting Pt:39…42 °C
- Density:1.07 g/cm³
- Flash Pt:79 °C
- Storage Temperature:Ambient
- EINECS:203-632-7
- UN:1671
- ADR:6.1,II
- REACH:01-2119471329-32
Documents
Specifications
Specification Test Results
- Appearance:Colorless to white Crystals or fused mass
- Infrared spectrum:Conforms
- Melting point:39 to 42 °C
- Water:≤0.2 %
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 kg



