Order Entry
Denmark
Orders LinkContactUsLinkComponent
n-Hexane 99+% for analysis

n-Hexane 99+% for analysis

Metal drum, 25 L
Catalog # ACRO160780251
Irritant
Health
Environment
Flammable
n-Hexane 99+% for analysis
Catalog # ACRO160780251
CAS Number:  110-54-3
n-Hexane 99+% for analysis
n-Hexane 99+% for analysis

Metal drum, 25 L

CAS Number:  110-54-3
Price
13.520,00 kr
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Boiling Pt:69 °C (760 mmHg)
  • Melting Pt:–95 °C
  • Density:0.659 g/cm³
  • Flash Pt:–22 °C
  • Storage Temperature:Ambient
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.02 % (Coulometric)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.002 %
  • Calcium (Ca):
    ≤0.5 ppm
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.1 ppm
  • Lead (Pb):
    ≤0.1 ppm
  • Magnesium (Mg):
    ≤0.1 ppm
  • Zinc (Zn):
    ≤0.1 ppm
  • Sulfur compounds (as S):
    ≤0.005 %
  • Free acid (CH3COOH):
    ≤0.002 %
  • Thiophene:
    Passes test

Specifications

  • Pack type:
    Metal drum
  • Pk:
    25 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.