D-(-)-Mannitol 98+%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:69-65-8
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064287
- Molecular Weight:182.17 g/mol
- Merck Index:13,05769
- Boiling Pt:290 - 295 °C (4 hPa)
- Melting Pt:165…170 °C
- Density:1,49 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:200-711-8
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:165 to 170 °C
- Loss on drying:≤0.3 % (105 °C, 4 hrs)
- HPLC:≥98.0 % (ELSD)
- Specific optical rotation:+23° to +25° (20 °C, 589 nm) (Ph.Eur.-directives)
- Nickel (Ni):≤1 ppm
- Heavy metals (as Pb):≤5 ppm
Specifications
- Pakning:Plastic bottle
- Pakn:100 g



