D-(-)-Mannitol 98+%
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:69-65-8
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064287
- Molecular Weight:182.17 g/mol
- Merck Index:13,05769
- Boiling Pt:290 - 295 °C (4 hPa)
- Melting Pt:165…170 °C
- Density:1,49 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-711-8
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:165 to 170 °C
- Loss on drying:≤0.3 % (105 °C, 4 hrs)
- HPLC:≥98.0 % (ELSD)
- Specific optical rotation:+23° to +25° (20 °C, 589 nm) (Ph.Eur.-directives)
- Nickel (Ni):≤1 ppm
- Heavy metals (as Pb):≤5 ppm
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



